3-Bromo-4-chlorobenzotrifluoride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2-chloro-5-(trifluoromethyl)benzene, 2-Bromo-1-chloro-4-(trifluoromethyl)benzene, 3-Bromo-4-chloro-α,α,αa-trifluorotoluene
PC1356-100G PC1356-250G PC1356-25G PC1356-500G
APOSPC1356-100GEA 43.5 EUR
APOSPC1356-100G APOSPC1356-250G APOSPC1356-25G APOSPC1356-500G
3-Bromo-4-chlorobenzotrifluoride 97%
3-Bromo-4-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
Boiling Pt: 188…192 °C
Melting Pt: –23 to –22 °C
Density: 1.726 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00018093
CAS Number: 454-78-4
EINECS: 207-226-0