1-Bromo-2,4-difluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Difluorobromobenzene
PC1382-100G PC1382-1KG PC1382-250G
APOSPC1382-100GEA 26.1 EUR
APOSPC1382-100G APOSPC1382-1KG APOSPC1382-250G
1-Bromo-2,4-difluorobenzene 98%
1-Bromo-2,4-difluorobenzene
Formula: C₆H₃BrF₂
Boiling Pt: 145…146 °C
Melting Pt: –4 °C
Density: 1.708 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000330
CAS Number: 348-57-2
EINECS: 206-479-4
UN: 1993
ADR: 3,III