1-Bromo-3,5-difluorobenzene 98%
Supplier: Apollo Scientific
Warning
PC1385-100G PC1385-1KG PC1385-250G
APOSPC1385-100GEA 21.9 EUR
APOSPC1385-100G APOSPC1385-1KG APOSPC1385-250G
1-Bromo-3,5-difluorobenzene 98%
1-Bromo-3,5-difluorobenzene
Formula: C₆H₃BrF₂
Boiling Pt: 140…141 °C
Melting Pt: –27 °C
Density: 1.676 g/cm³ (25 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00010305
CAS Number: 461-96-1
UN: 1993
ADR: 3,III