3-Bromo-α,α,α,2-tetrafluorotoluene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-2-fluorobenzotrifluoride, 1-Bromo-2-fluoro-3-(trifluoromethyl)benzene
PC1421M-5G PC1421M-100G PC1421M-25G
APOSPC1421M-5GEA 27.7 EUR
APOSPC1421M-5G APOSPC1421M-100G APOSPC1421M-25G
3-Bromo-α,α,α,2-tetrafluorotoluene 97%
3-Bromo-α,α,α,2-tetrafluorotoluene
Formula: C₇H₃BrF₄
Density: 1.72 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00070812
CAS Number: 144584-67-8