2-Bromo-4-fluorophenol 97%
Supplier: Apollo Scientific
Warning
PC1429-100G PC1429-500G PC1429-25G
APOSPC1429-100GEA 49.3 EUR
APOSPC1429-100G APOSPC1429-500G APOSPC1429-25G
2-Bromo-4-fluorophenol 97%
2-Bromo-4-fluorophenol
Formula: C₆H₄BrFO
Boiling Pt: 130…131 °C/50mmHg
Melting Pt: 42…44 °C
Density: 1.717 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00010614
CAS Number: 496-69-5