2-Bromo-4-fluorotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3-fluoro-6-methylbenzene
PC1430-100G PC1430-500G PC1430-1KG
APOSPC1430-100GEA 24.8 EUR
APOSPC1430-100G APOSPC1430-500G APOSPC1430-1KG
2-Bromo-4-fluorotoluene 98%
2-Bromo-4-fluorotoluene
Formula: C₇H₆BrF
Boiling Pt: 170…172 °C
Density: 1.526 g/cm³ (25 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00040828
CAS Number: 1422-53-3
EINECS: 215-830-0