Ethyl-8-fluoro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indole-2-carboxylate 95%
Supplier: Apollo Scientific
PC1445-10G PC1445-5G PC1445-1G
APOSPC1445-10GEA 894 EUR
APOSPC1445-10G APOSPC1445-5G APOSPC1445-1G
Ethyl-8-fluoro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indole-2-carboxylate 95%
Ethyl-8-fluoro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indole-2-carboxylate
Formula: C₁₄H₁₅FN₂O₂
Melting Pt: 172…173 °C
Storage Temperature: Ambient
MDL Number: MFCD02930345
CAS Number: 58038-66-7