4-Bromo-2-fluorotoluene 99%
Supplier: Apollo Scientific
Warning
PC1450-100G PC1450-1KG PC1450-25G PC1450-500G
APOSPC1450-100GEA 89.9 EUR
APOSPC1450-100G APOSPC1450-1KG APOSPC1450-25G APOSPC1450-500G
4-Bromo-2-fluorotoluene 99%
4-Bromo-2-fluorotoluene
Formula: C₇H₆BrF
Boiling Pt: 68 °C/8mmHg
Density: 1.492 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00013551
CAS Number: 51436-99-8
EINECS: 257-201-3