Ethyl 1-(4-fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylate 95%
Supplier: Apollo Scientific
Warning
PC1453-1G PC1453-500MG PC1453-5G
APOSPC1453-1GEA 275 EUR
APOSPC1453-1G APOSPC1453-500MG APOSPC1453-5G
Ethyl 1-(4-fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylate 95%
Ethyl-1-(4-fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylate
Formula: C₁₃H₁₄FN₃O₂
Melting Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD06797391
CAS Number: 885950-26-5