3-Bromo-5-fluoroanisole 96%
Supplier: Apollo Scientific
Warning
PC1483-100G PC1483-25G PC1483-5G
APOSPC1483-100GEA 95.7 EUR
APOSPC1483-100G APOSPC1483-25G APOSPC1483-5G
3-Bromo-5-fluoroanisole 96%
3-Bromo-5-fluoroanisole
Formula: C₇H₆BrFO
Boiling Pt: 94 °C/17mmHg
Density: 1.59 g/cm³ (25 °C)
Flash Pt: >90 °C
Storage Temperature: Ambient
MDL Number: MFCD01861129
CAS Number: 29578-39-0