2,3,6-Trifluorophenylboronic acid 98%
Supplier: Apollo Scientific
Warning
PC1512-25G PC1512-5G PC1512-100G
APOSPC1512-25GEA 104 EUR
APOSPC1512-25G APOSPC1512-5G APOSPC1512-100G
2,3,6-Trifluorophenylboronic acid 98%
2,3,6-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
Melting Pt: 127…132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863167
CAS Number: 247564-71-2