1-Bromo-2,3,4-trifluorobenzene 98%
Supplier: Apollo Scientific
Warning
PC1568-100G PC1568-250G PC1568-25G
APOSPC1568-100GEA 79.9 EUR
APOSPC1568-100G APOSPC1568-250G APOSPC1568-25G
1-Bromo-2,3,4-trifluorobenzene 98%
1-Bromo-2,3,4-trifluorobenzene
Formula: C₆H₂BrF₃
Boiling Pt: 47…49 °C/60mmHg
Density: 1.777 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00013248
CAS Number: 176317-02-5