4'-Bromo-3'-(trifluoromethyl)acetanilide 98%
Supplier: Apollo Scientific
Warning
Synonyms: N-(4-Bromo-3-(trifluoromethyl)phenyl)acetamide
PC1594-1G PC1594-5G
APOSPC1594-1GEA 21.9 EUR
APOSPC1594-1G APOSPC1594-5G
4'-Bromo-3'-(trifluoromethyl)acetanilide 98%
4'-Bromo-3'-(trifluoromethyl)acetanilide
Formula: C₉H₇BrF₃NO
MW: 282.07 g/mol
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00052361
CAS Number: 41513-05-7