3-Bromo-2-fluoroanisole 98+%
Supplier: Apollo Scientific
Warning
PC1688-10G PC1688-1G PC1688-5G
APOSPC1688-10GEA 72.5 EUR
APOSPC1688-10G APOSPC1688-1G APOSPC1688-5G
3-Bromo-2-fluoroanisole 98+%
3-Bromo-2-fluoroanisole
Formula: C₇H₆BrFO
Boiling Pt: 207 °C
Density: 1.531 g/cm³ (25 °C)
Flash Pt: 93.4 °C
Storage Temperature: Ambient
MDL Number: MFCD07780183
CAS Number: 295376-21-5