2,3'-Dichloro-4'-fluoroacetanilide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloro-N-(chloroacetyl)-4-fluoroaniline
PC1702-1G PC1702-500MG
APOSPC1702-1GEA 197 EUR
APOSPC1702-1G APOSPC1702-500MG
2,3'-Dichloro-4'-fluoroacetanilide 97%
2,3'-Dichloro-4'-fluoroacetanilide
Formula: C₈H₆Cl₂FNO
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00084938
CAS Number: 96980-64-2