2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid 98%
Supplier: Apollo Scientific
Synonyms: (2-Isopropoxy-5-(trifluoromethyl)phenyl)boronic acid, 2-Isopropoxy-5-(trifluoromethyl)benzeneboronic acid, 2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid
PC1775-1G PC1775-5G
APOSPC1775-1GEA 74.4 EUR
APOSPC1775-1G APOSPC1775-5G
2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid 98%
2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid
Formula: C₁₀H₁₂BF₃O₃
MW: 248.0100 g/mol
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD07363786
CAS Number: 850593-12-3