8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Supplier: Apollo Scientific
PC1931K-1G PC1931K-5G
APOSPC1931K-1GEA 35 EUR
APOSPC1931K-1G APOSPC1931K-5G
8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃NO
MW: 247.61 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00276589
CAS Number: 18706-22-4