3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine 97+%
Supplier: Apollo Scientific
Warning
PC1936E-1G PC1936E-5G
APOSPC1936E-1GEA 21.9 EUR
APOSPC1936E-1G APOSPC1936E-5G
3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine 97+%
3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine
Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
Boiling Pt: 173…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00153096
CAS Number: 175136-17-1