2-Chloro-5-nitrobenzotrifluoride 98%
Supplier: Apollo Scientific
Warning
PC1940-100G PC1940-25G PC1940-500G
APOSPC1940-100GEA 24.8 EUR
APOSPC1940-100G APOSPC1940-25G APOSPC1940-500G
2-Chloro-5-nitrobenzotrifluoride 98%
2-Chloro-5-nitrobenzotrifluoride
Formula: C₇H₃ClF₃NO₂
Boiling Pt: 102…103 °C/5mmHg
Melting Pt: 22 °C
Density: 1.527 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007296
CAS Number: 777-37-7
EINECS: 212-287-1