1-Chloro-4H-octafluorobutane 97%

Supplier: Apollo Scientific
Warning

PC1965-1G PC1965-25G PC1965-5G
APOSPC1965-1GEA 157 EUR
APOSPC1965-1G APOSPC1965-25G APOSPC1965-5G
1-Chloro-4H-octafluorobutane 97%
1-Chloro-4H-octafluorobutane
Formula: C₄HClF₈
Storage Temperature: Ambient
MDL Number: MFCD00155682
CAS Number: 423-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR