2-Amino-3,5-difluoro-4-(trifluoromethyl)pyridine 95%
Supplier: Apollo Scientific
Warning
PC1991-1G PC1991-250MG
APOSPC1991-1GEA 357 EUR
APOSPC1991-1G APOSPC1991-250MG
2-Amino-3,5-difluoro-4-(trifluoromethyl)pyridine 95%
2-Amino-3,5-difluoro-4-(trifluoromethyl)pyridine
Formula: C₆H₃F₅N₂
Boiling Pt: 85…86 °C/27mmHg
Density: 1.524 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03840199
CAS Number: 883498-68-8