1-[(4-Fluorophenyl)methyl]-1H-indazole-3-carboxylic acid
Supplier: Apollo Scientific
PC200018-1G PC200018-500MG
APOSPC200018-1GEA 538 EUR
APOSPC200018-1G APOSPC200018-500MG
1-[(4-Fluorophenyl)methyl]-1H-indazole-3-carboxylic acid
1-[(4-Fluorophenyl)methyl]-1H-indazole-3-carboxylic acid
Formula: C₁₅H₁₁FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01719717
CAS Number: 50264-63-6