2-Chloro-3-iodo-4-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
PC200082-1G PC200082-500MG PC200082-5G
APOSPC200082-1GEA 470 EUR
APOSPC200082-1G APOSPC200082-500MG APOSPC200082-5G
2-Chloro-3-iodo-4-(trifluoromethyl)pyridine
2-Chloro-3-iodo-4-(trifluoromethyl)pyridine
Formula: C₆H₂ClF₃IN
MW: 307.44 g/mol
Melting Pt: 55…56 °C
Storage Temperature: Ambient
MDL Number: MFCD12827836
CAS Number: 1186405-09-3