2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1,2-oxazolidin-4-ol
Supplier: Apollo Scientific
PC200163-1G PC200163-500MG
APOSPC200163-1GEA 210 EUR
APOSPC200163-1G APOSPC200163-500MG
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1,2-oxazolidin-4-ol
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1,2-oxazolidin-4-ol
Formula: C₉H₈ClF₃N₂O₂
MW: 268.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21333205
CAS Number: 1357147-43-3