(Z)-2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(dimethylamino)-2-propenenitrile
Supplier: Apollo Scientific
PC200168-1G PC200168-500MG
APOSPC200168-1GEA 336 EUR
APOSPC200168-1G APOSPC200168-500MG
(Z)-2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(dimethylamino)-2-propenenitrile
(Z)-2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(dimethylamino)-2-propenenitrile
Formula: C₁₁H₉ClF₃N₃
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD16622821
CAS Number: 1242271-46-0