7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
PC200218-1G PC200218-500MG
APOSPC200218-1GEA 415 EUR
APOSPC200218-1G APOSPC200218-500MG
7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Formula: C₈H₃ClF₃IN₂
MW: 346.48 g/mol
Melting Pt: 160…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19982794
CAS Number: 1331823-98-3