3-(Trifluoromethyl)quinoxaline-2-carboxylic acid
Supplier: Apollo Scientific
Warning
PC200243-1G PC200243-500MG
APOSPC200243-1GEA 415 EUR
APOSPC200243-1G APOSPC200243-500MG
3-(Trifluoromethyl)quinoxaline-2-carboxylic acid
3-(Trifluoromethyl)quinoxaline-2-carboxylic acid
Formula: C₁₀H₅F₃N₂O₂
Melting Pt: 164 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD11869266
CAS Number: 1142190-60-0