Methyl-2-[8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate
Supplier: Apollo Scientific
Warning
PC200248-1G PC200248-500MG
APOSPC200248-1GEA 415 EUR
APOSPC200248-1G APOSPC200248-500MG
Methyl-2-[8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate
Methyl-2-[8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate
Formula: C₁₁H₈F₃IN₂O₂
MW: 384.1 g/mol
Melting Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11553037
CAS Number: 1092352-47-0