2,3-Dichloro-5-(trifluoromethyl)isonicotinonitrile
Supplier: Apollo Scientific
Warning
PC200264-1G PC200264-500MG
APOSPC200264-1GEA 633 EUR
APOSPC200264-1G APOSPC200264-500MG
2,3-Dichloro-5-(trifluoromethyl)isonicotinonitrile
2,3-Dichloro-5-(trifluoromethyl)isonicotinonitrile
Formula: C₇HCl₂F₃N₂
Melting Pt: 51…52 °C
Storage Temperature: Ambient
MDL Number: MFCD14581647
CAS Number: 1198475-29-4