3-Iodo-5-methoxy-6-(trifluoromethyl)-1H-indole
Supplier: Apollo Scientific
PC200276-1G PC200276-500MG
APOSPC200276-1GEA 519 EUR
APOSPC200276-1G APOSPC200276-500MG
3-Iodo-5-methoxy-6-(trifluoromethyl)-1H-indole
3-Iodo-5-methoxy-6-(trifluoromethyl)-1H-indole
Formula: C₁₀H₇F₃INO
MW: 341.07 g/mol
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD12498701
CAS Number: 1171918-93-6