4-Chloro-2-(trifluoromethyl)quinazoline
Supplier: Apollo Scientific
Warning
PC200285-1G PC200285-500MG PC200285-5G
APOSPC200285-1GEA 295 EUR
APOSPC200285-1G APOSPC200285-500MG APOSPC200285-5G
4-Chloro-2-(trifluoromethyl)quinazoline
4-Chloro-2-(trifluoromethyl)quinazoline
Formula: C₉H₄ClF₃N₂
Melting Pt: 65…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00793683
CAS Number: 52353-35-2