5-Bromo-6-(trifluoromethyl)-1H-indole
Supplier: Apollo Scientific
Warning
PC200287-1G PC200287-500MG
APOSPC200287-1GEA 720 EUR
APOSPC200287-1G APOSPC200287-500MG
5-Bromo-6-(trifluoromethyl)-1H-indole
5-Bromo-6-(trifluoromethyl)-1H-indole
Formula: C₉H₅BrF₃N
Melting Pt: 66…68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094968
CAS Number: 1198475-24-9