Ethyl-5,7-bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylate
Supplier: Apollo Scientific
PC200349-1G PC200349-500MG
APOSPC200349-1GEA 487 EUR
APOSPC200349-1G APOSPC200349-500MG
Ethyl-5,7-bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylate
Ethyl-5,7-bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylate
Formula: C₁₃H₈F₆N₂O₂
MW: 338.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22373660
CAS Number: 1379526-96-1