5,7-Bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylic acid
Supplier: Apollo Scientific
PC200352-1G PC200352-500MG
APOSPC200352-1GEA 611 EUR
APOSPC200352-1G APOSPC200352-500MG
5,7-Bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylic acid
5,7-Bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylic acid
Formula: C₁₁H₄F₆N₂O₂
MW: 310.16 g/mol
Melting Pt: 171 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD22373654
CAS Number: 1379526-94-9