4-Chloro-3-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Chloro-5-cyanobenzotrifluoride
PC2004-1G PC2004-25G PC2004-5G PC2004-100G
APOSPC2004-1GEA 14.7 EUR
APOSPC2004-1G APOSPC2004-25G APOSPC2004-5G APOSPC2004-100G
4-Chloro-3-(trifluoromethyl)benzonitrile 98%
4-Chloro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
MW: 205.57
Boiling Pt: 100…105 °C/11mmHg
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00190117
CAS Number: 1735-54-2
UN: 3439
ADR: 6.1 III