4-Chloro-2,3-difluoroaniline 98%
Supplier: Apollo Scientific
PC201342-10G PC201342-1G PC201342-5G
APOSPC201342-10GEA 424 EUR
APOSPC201342-10G APOSPC201342-1G APOSPC201342-5G
4-Chloro-2,3-difluoroaniline 98%
4-Chloro-2,3-difluoroaniline
Formula: C₆H₄ClF₂N
MW: 163.5500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08459284
CAS Number: 878285-12-2