3-Chloro-2,2,3,3-tetrafluoropropionic acid 97%

Supplier: Apollo Scientific
Danger

PC2026G-1G PC2026G-5G
APOSPC2026G-1GEA 29.2 EUR
APOSPC2026G-1G APOSPC2026G-5G
3-Chloro-2,2,3,3-tetrafluoropropionic acid 97%
3-Chloro-2,2,3,3-tetrafluoropropionic acid
Formula: C₃HClF₄O₂
Boiling Pt: 130…131 °C
Density: 1.63 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00153099
CAS Number: 661-82-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR