2-Chloro-1,1,2-trifluoro-1-methoxyethane 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Chloro-1,1,2-trifluoroethyl methyll ether
PC2070-100G PC2070-25G
APOSPC2070-100GEA 760 EUR
APOSPC2070-100G APOSPC2070-25G
2-Chloro-1,1,2-trifluoro-1-methoxyethane 97%
2-Chloro-1,1,2-trifluoro-1-methoxyethane
Formula: C₃H₄ClF₃O
MW: 148.51 g/mol
Boiling Pt: 69…71.5 °C
Melting Pt: –92 °C
Density: 1.363 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00039335
CAS Number: 425-87-6
EINECS: 207-038-9
UN: 2810
ADR: 6.1 III