4-Chloro-6-(trifluoromethyl)-1H-benzimidazole 97%
Supplier: Apollo Scientific
Warning
PC2073R-1G PC2073R-500MG PC2073R-5G
APOSPC2073R-1GEA 109 EUR
APOSPC2073R-1G APOSPC2073R-500MG APOSPC2073R-5G
4-Chloro-6-(trifluoromethyl)-1H-benzimidazole 97%
4-Chloro-6-(trifluoromethyl)-1H-benzimidazole
Formula: C₈H₄ClF₃N₂
MW: 220.58 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00067733
CAS Number: 175135-13-4