N-[2-Chloro-5-(trifluoromethyl)phenyl]thiourea
Supplier: Apollo Scientific
PC2074T-1G PC2074T-5G PC2074T-10G
APOSPC2074T-1GEA 65.4 EUR
APOSPC2074T-1G APOSPC2074T-5G APOSPC2074T-10G
N-[2-Chloro-5-(trifluoromethyl)phenyl]thiourea
N-[2-Chloro-5-(trifluoromethyl)phenyl]thiourea
Formula: C₈H₆ClF₃N₂S
MW: 254.66 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00041155
CAS Number: 21714-35-2