4'-Chloro-3'-(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-3-(trifluoromethyl)acetophenone
PC2172-100G PC2172-25G
APOSPC2172-100GEA 178 EUR
APOSPC2172-100G APOSPC2172-25G
4'-Chloro-3'-(trifluoromethyl)acetophenone
4'-Chloro-3'-(trifluoromethyl)acetophenone
Formula: C₉H₆ClF₃O
MW: 222.59 g/mol
Melting Pt: 61…63 °C
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD03094146
CAS Number: 129825-11-2