1,3-Dibromo-1,1-difluorobutane 97%
Supplier: Apollo Scientific
PC2237F-25G PC2237F-5G
APOSPC2237F-25GEA 789 EUR
APOSPC2237F-25G APOSPC2237F-5G
1,3-Dibromo-1,1-difluorobutane 97%
1,3-Dibromo-1,1-difluorobutane
Formula: C₄H₆Br₂F₂
Boiling Pt: 60…61 °C/50mmHg
Melting Pt: 62…64 °C
Storage Temperature: Ambient
CAS Number: 406-42-8