1,4-Dibromooctafluorobutane 98%
Supplier: Apollo Scientific
Warning
PC2279G-25G PC2279G-5G PC2279G-100G
APOSPC2279G-25GEA 132 EUR
APOSPC2279G-25G APOSPC2279G-5G APOSPC2279G-100G
1,4-Dibromooctafluorobutane 98%
1,4-Dibromooctafluorobutane
Formula: C₄Br₂F₈
Boiling Pt: 96…98 °C
Density: 2.0979 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00153112
CAS Number: 335-48-8