2,3-Dichloro-α,α,α-trifluorotoluene 98%
Supplier: Apollo Scientific
Warning
PC2334-5G PC2334-25G
APOSPC2334-5GEA 21.9 EUR
APOSPC2334-5G APOSPC2334-25G
2,3-Dichloro-α,α,α-trifluorotoluene 98%
2,3-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Melting Pt: 18…19 °C
Density: 1.52 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00069115
CAS Number: 54773-19-2
EINECS: 259-336-3