2-Chloro-4-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Danger
PC2437-100G PC2437-25G PC2437-5G
APOSPC2437-100GEA 295 EUR
APOSPC2437-100G APOSPC2437-25G APOSPC2437-5G
2-Chloro-4-(trifluoromethyl)pyridine 98%
2-Chloro-4-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 145…147 °C
Melting Pt: -1
Density: 1.411 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00042224
CAS Number: 81565-18-6
UN: 1992
ADR: 3+6.1 III