3-Bromo-2-fluorotoluene 98%
Supplier: Apollo Scientific
Warning
PC2446-100G PC2446-25G PC2446-500G
APOSPC2446-100GEA 84.1 EUR
APOSPC2446-100G APOSPC2446-25G APOSPC2446-500G
3-Bromo-2-fluorotoluene 98%
3-Bromo-2-fluorotoluene
Formula: C₇H₆BrF
Boiling Pt: 185…186 °C/760mmHg
Density: 1.503 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD04972694
CAS Number: 59907-12-9
EINECS: 261-981-0