(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate 98%
Supplier: Apollo Scientific
Warning
Synonyms: (1-(Ethoxycarbonyl)cyclopropyl)triphenylphosphonium tetrafluoroborate
PC2522-25G PC2522-1G PC2522-5G
APOSPC2522-25GEA 724 EUR
APOSPC2522-25G APOSPC2522-1G APOSPC2522-5G
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate 98%
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate
Fuchs Reagent
Formula: C₂₄H₂₄BF₄O₂P
MW: 462.23 g/mol
Melting Pt: 173…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00051879
CAS Number: 52186-89-7