4-Bromo-2,3-difluorophenol
Supplier: Apollo Scientific
Warning
PC2553-5G PC2553-10G PC2553-1G
APOSPC2553-5GEA 24.8 EUR
APOSPC2553-5G APOSPC2553-10G APOSPC2553-1G
4-Bromo-2,3-difluorophenol
4-Bromo-2,3-difluorophenol
Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD08061907
CAS Number: 144292-32-0