3,4-Difluorobenzaldehyde 96%
Supplier: Apollo Scientific
Warning
PC2615E-100G PC2615E-10G PC2615E-25G
APOSPC2615E-100GEA 85.7 EUR
APOSPC2615E-100G APOSPC2615E-10G APOSPC2615E-25G
3,4-Difluorobenzaldehyde 96%
3,4-Difluorobenzaldehyde
Formula: C₇H₄F₂O
Boiling Pt: 180 °C
Density: 1.288 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00010328
CAS Number: 34036-07-2
EINECS: 422-180-3