2,5-Difluorobenzamide 97%
Supplier: Apollo Scientific
Warning
PC2617-1G PC2617-5G PC2617-10G PC2617-25G
APOSPC2617-1GEA 14.7 EUR
APOSPC2617-1G APOSPC2617-5G APOSPC2617-10G APOSPC2617-25G
2,5-Difluorobenzamide 97%
2,5-Difluorobenzamide
Formula: C₇H₅F₂NO
MW: 157.12 g/mol
Melting Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00015548
CAS Number: 85118-03-2